ContactUsLinkComponent
Reference guide and photographs for steel surfaces, SSPC-VIS 4/NACE VIS 7
Reference guide and photographs for steel surfaces, SSPC-VIS 4/NACE VIS 7
Catalog # SIMELD3058
Supplier:  Industrial Physics
undefined
Reference guide and photographs for steel surfaces, SSPC-VIS 4/NACE VIS 7
Catalog # SIMELD3058
Supplier:  Industrial Physics
Specifications
  • Description:
    Guide and reference photographs for steel surfaces prepared by waterjetting
  • Pk:
    1
Specifications
About this item
The SSPC-VIS 4/NACE VIS 7 shows six different steel surfaces - two uncoated and four previously coated - before and after waterjetting.
  • Photographs illustrate four separate degrees of cleaning (WJ 1, WJ 2, WJ 3, and WJ 4) for each initial condition
  • Additional photos that depict the appearance light, moderate, and heavy flash rust after cleaning
  • Cleanliness requirements for conditions depicted are defined
  • Joint SSPC/NACE surface preparation specification for high and ultra high pressure water jetting (SP 12/NACE 5)
Ordering information: Contains a written guide to the use of reference photographs and additional explanatory notes.