Order Entry
Czech Republic
ContactUsLinkComponent
 
Acrylamide : Bisacrylamide (37.5:1) 40% in aqueous solution, Liqui-Gel™
  ICNA04800804
 :  
Acrylamide : Bisacrylamide (37.5:1) 40% in aqueous solution, Liqui-Gel™
  ICNA04800804
 :  

 

  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Form:
    Clear, Colorless solution
  • Storage:
    Store at Room Temperature (15 - 30 °C)
  • Feed Ratio:
    37:5:1

 

 

 :
 :79-06-1
 :MFCD00080848