Anti-KDM5B Rabbit Polyclonal Antibody
BSBTPB10073
- Antibody type:Primary
- Antigen name:Lysine (K)-Specific Demethylase 5B
- Antigen symbol:KDM5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10765
- Antigen synonyms:PLU1|JARID1B|CT31|RBP2-H1|PUT1|RBBP2H1A|PPP1R98|PLU-1
- Size:100 μg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Lysine-specific demethylase 5B(KDM5B) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: KDM5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat