Anti-BDKRB2 Rabbit Polyclonal Antibody
Catalog # BSBTPB10047
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Bradykinin Receptor B2
- Antigen symbol:BDKRB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:624
- Antigen synonyms:BKR2|BK2|BRB2|BK-2|B2R
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for B2 bradykinin receptor(BDKRB2) detection. Tested with WB in Human.
Type: Primary
Antigen: BDKRB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human