Anti-AOC1 Rabbit Polyclonal Antibody
Catalog # BSBTPB10040
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Amine Oxidase, Copper Containing 1
- Antigen symbol:AOC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:26
- Antigen synonyms:ABP1|DAO|KAO|DAO1|ABP
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Amiloride-sensitive amine oxidase [copper-containing](AOC1) detection. Tested with WB in Human;Rat.
Type: Primary
Antigen: AOC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat