Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-ITGB14 Mouse Monoclonal Antibody (Biotin) [clone: VJ1/14]
  ABNOMAB13866
 :  Abnova
Anti-ITGB14 Mouse Monoclonal Antibody (Biotin) [clone: VJ1/14]
  ABNOMAB13866
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen symbol:
    ITGB14
  • Clonality:
    Monoclonal
  • Clone:
    VJ1/14
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Gene ID:
    3688
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Mouse monoclonal antibody raised against human ITGB1.

Type: Primary
Antigen: ITGB14
Clonality: Monoclonal
Clone: VJ1/14
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human