Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ITGAX Mouse Monoclonal Antibody (Biotin) [clone: BU-15]
Anti-ITGAX Mouse Monoclonal Antibody (Biotin) [clone: BU-15]
  ABNOMAB13803
 :  Abnova
Anti-ITGAX Mouse Monoclonal Antibody (Biotin) [clone: BU-15]
  ABNOMAB13803
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen symbol:
    ITGAX
  • Clonality:
    Monoclonal
  • Clone:
    BU-15
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Gene ID:
    3687
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Mouse monoclonal antibody raised against human ITGAX.

Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: BU-15
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human