Anti-ITGAX Mouse Monoclonal Antibody (PerCP) [clone: BU-15]
ABNOMAB13802
: Abnova
- Antibody type:Primary
- Antigen symbol:ITGAX
- Clonality:Monoclonal
- Clone:BU-15
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:3687
- Size:100 reactions
- Pk:100 Assays
Mouse monoclonal antibody raised against human ITGAX.
Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: BU-15
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human