Anti-ITGAM Mouse Monoclonal Antibody (PerCP) [clone: DCIS1/18]
Catalog # ABNOMAB13799
Dodavatel: Abnova
Specifikace
- Antibody type:Primary
- Antigen symbol:ITGAM
- Clonality:Monoclonal
- Clone:DCIS1/18
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Gene ID:3684
- Size:100 reactions
- Pk:100 Assays
Specifikace
O tomto produktu
Mouse monoclonal antibody raised against human ITGAM.
Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: DCIS1/18
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human