Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ITGAM Mouse Monoclonal Antibody (PE) [clone: DCIS1/18]
Anti-ITGAM Mouse Monoclonal Antibody (PE) [clone: DCIS1/18]
Catalog # ABNOMAB13797
Supplier:  Abnova
Anti-ITGAM Mouse Monoclonal Antibody (PE) [clone: DCIS1/18]
Catalog # ABNOMAB13797
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ITGAM
  • Clonality:
    Monoclonal
  • Clone:
    DCIS1/18
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Gene ID:
    3684
  • Size:
    100 reactions
  • Pk:
    100 Assays

Specifications

About this item

Mouse monoclonal antibody raised against human ITGAM.

Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: DCIS1/18
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human