Anti-CD146 Mouse Monoclonal Antibody (PerCP) [clone: 47-3D6]
Catalog # ABNOMAB13807
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:CD146
- Clonality:Monoclonal
- Clone:47-3D6
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Gene ID:929
- Size:100 reactions
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human CD14.
Type: Primary
Antigen: CD146
Clonality: Monoclonal
Clone: 47-3D6
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human,Mouse