Anti-PECAM15 Mouse Monoclonal Antibody (Biotin) [clone: TP1/15]
Catalog # ABNOMAB13872
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:PECAM15
- Clonality:Monoclonal
- Clone:TP1/15
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Gene ID:5175
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against native human PECAM1.
Type: Primary
Antigen: PECAM15
Clonality: Monoclonal
Clone: TP1/15
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human