Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ITGA241 Mouse Monoclonal Antibody (FITC) [clone: TEA1/41]
Anti-ITGA241 Mouse Monoclonal Antibody (FITC) [clone: TEA1/41]
Catalog # ABNOMAB13688
Supplier:  Abnova
Anti-ITGA241 Mouse Monoclonal Antibody (FITC) [clone: TEA1/41]
Catalog # ABNOMAB13688
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ITGA241
  • Clonality:
    Monoclonal
  • Clone:
    TEA1/41
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Gene ID:
    3673
  • Size:
    100 reactions
  • Pk:
    100 Assays

Specifications

About this item

Mouse monoclonal antibody raised against human ITGA2.

Type: Primary
Antigen: ITGA241
Clonality: Monoclonal
Clone: TEA1/41
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human