Anti-EPCAM1 Mouse Monoclonal Antibody [clone: MOC-31]
Catalog # ABNOMAB13331
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:EPCAM1
- Clonality:Monoclonal
- Clone:MOC-31
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:4072
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against native human EPCAM.
Type: Primary
Antigen: EPCAM1
Clonality: Monoclonal
Clone: MOC-31
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human