Anti-Enterococcus species Rabbit Polyclonal Antibody
Catalog # ABNOPAB29839
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Enterococcus species
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Bacteria
- Size:500 μL
- Pk:500 µl
Specifications
About this item
Rabbit polyclonal antibody raised against Enterococcus species.
Type: Primary
Antigen: Enterococcus species
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria