Order Entry
Czech Republic
ContactUsLinkComponent
VWR® VividPro™ Fluorescent Stains
VWR® VividPro™ Fluorescent Stains
Supplier:  VWR Chemicals
Certificates
undefined
VWR® VividPro™ Fluorescent Stains
Supplier:  VWR Chemicals

Specifications

  • Excitation:
    284, 366 nm (0,2 mg/ml BSA)
  • Emission:
    471 nm (0,2 mg/ml BSA)
  • Sensitivity:
    10 ng per protein band

Specifications

Related Information

Frequently Bought Together