ContactUsLinkComponent
Wall bracket for eyewash bottles
Wall bracket for eyewash bottles
Catalog # CEDE7202
Supplier:  ORKLA CARE
undefined
Wall bracket for eyewash bottles
Catalog # CEDE7202
Supplier:  ORKLA CARE
Specifications
  • Description:
    Wall bracket for 2 Cederroth eyewash bottles (500 ml), incl. afterglowing sign, 293×170×85 mm (wall bracket), 85×250 mm (sign)
  • Pk:
    1
Specifications
About this item
Two holders in green hard plastics intended for storage of two Cederroth eyewash bottles (500 ml), making it easy to find in an emergency situation.
Holders are fastened on a metal plate, which can be mounted on any vertical surface example on a wall.
Also serves as an opening device for the eyewash bottles: Bottle is automatically opened when removed from the holder.
Afterglowing sign with instructions for use is included.