Universal valve lockout, base clamping units
: Brady
These universal valve lockouts are effective for all types of standard valves.
- Versatile, rugged, easy to use and lightweight
- Can also be used to lock out large levers, T-handles and hard-to-secure mechanical devices
- Open-ended clamp fits over closed rings and wide handles
- Made of industrial-grade steel and nylon for extra impact and chemical resistance
Max. handle thickness: 15,24 mm
Max. valve handle width: 25,40 mm
Max. shackle diameter: 9,00 mm
Min. shackle clearance: 20,00 mm
: Base unit only (cable and blocking arm sold separately).
Frequently Bought Together