Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD86 Rat Monoclonal Antibody (PE-Cyanine7) [clone: GL-1 (GL1)]
Anti-CD86 Rat Monoclonal Antibody (PE-Cyanine7) [clone: GL-1 (GL1)]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD86 Rat Monoclonal Antibody (PE-Cyanine7) [clone: GL-1 (GL1)]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 60-0862-U025
  • 60-0862-U100
  • Antigen symbol:
  • CD86
  • CD86
  • Antigen name:
  • CD86, CD28 ligand
  • CD86, CD28 ligand
  • Antigen synonyms:
  • B70|B7.2|Ly-58
  • B70|B7.2|Ly-58
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)-Cy7®
  • PE (Phycoerythrin)-Cy7®
  • Clone:
  • GL-1 (GL1)
  • GL-1 (GL1)
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12524
  • 12524
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • +2 °C to +8 °C, protected from light
  • +2 °C to +8 °C, protected from light

Specifications

Related Information

Frequently Bought Together