Mouse IgG2b Isotype Control [clone: MPC-11] (APC (Allophycocyanin)-Cy7®)
Catalog # 25-4732-U100
Dodavatel: Tonbo Biosciences
Specifikace
- Antibody type:Control
- Clonality:Monoclonal
- Clone:MPC-11
- Conjugation:APC (Allophycocyanin)-Cy7®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Storage temperature:+2 °C to +8 °C, protected from light
- Size:100 μg
- Pk:100 µG
Specifikace
O tomto produktu
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.
Type: Control
Antigen: Isotype Control
Clonality: Monoclonal
Clone: MPC-11
Conjugation: APC-Cyanine7
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: N/A