The MEL-14 antibody is specific for mouse CD62L, also known as L-Selectin, a cell adhesion molecule which facilitates lymphocyte "rolling" on activated vascular endothelium and homing to high endothelial venules (HEV) as immune cells transmigrate from blood into peripheral tissues. L-Selectin is a member of a family of Selectin molecules which act together with the integrin family of adhesion molecules to mediate leukocyte-endothelial interactions. L-Selectin is characteristically expressed by neutrophils, and is also found on B cells, monocytes, granulocytes, and at varying levels on naive, effector and memory T cells. It is rapidly shed upon cell activation, releasing into the circulation a soluble form whose biological role is of particular interest in cancer biology research. The MEL-14 antibody may be used as a phenotypic marker for CD62L expression on a variety of immune cell types. Please note that CD62L (L-Selectin) itself is also referred to as MEL-14 in the literature.
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: MEL-14
Conjugation: APC-Cyanine7
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
- Catalog No:
- 25-0621-U025
- 25-0621-U100
- Antigen symbol:
- CD62L
- CD62L
- Antigen name:
- CD62L, L-Selectin
- CD62L, L-Selectin
- Antigen synonyms:
- Ly-22|LECAM-1
- Ly-22|LECAM-1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)-Cy7®
- APC (Allophycocyanin)-Cy7®
- Clone:
- MEL-14
- MEL-14
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 20343
- 20343
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- +2 °C to +8 °C, protected from light
- +2 °C to +8 °C, protected from light