About this item
The 145-2C11 antibody is specific for mouse CD3e, also known as CD3 epsilon, a 20 kDa subunit of the T cell receptor complex, along with CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. Such interactions promote T cell receptor signaling (T cell activation) and can result in a number of cellular responses including proliferation, differentiation, production of cytokines or activation-induced cell death. CD3 is differentially expressed during thymocyte-to-T cell development and on all mature T cells. The 145-2C11 antibody is a widely used phenotypic marker for mouse T cells. In addition, binding of 145-2C11 antibody to CD3e can induce cell activation. A recent publication of the crystal structure of a murine CD3e-mitogenic antibody complex provides further insight into the action of commonly used agonist antibodies (Fernandes, R.A. et al. 2012. J. Biol. Chem. 287: 13324-13335).
Type: Primary
Antigen: CD3e
Clonality:
Clone: 145-2C11
Conjugation: APC-Cyanine7
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 25-0031-U100
- 25-0031-U025
- Antigen symbol:
- CD3e
- CD3e
- Antigen name:
- CD3 epsilon subunitT cell receptor complex
- CD3 epsilon subunitT cell receptor complex
- Antigen synonyms:
- CD3 epsilon
- CD3 epsilon
- Antibody type:
- Primary
- Primary
- Conjugation:
- APC (Allophycocyanin)-Cy7®
- APC (Allophycocyanin)-Cy7®
- Clone:
- 145-2C11
- 145-2C11
- Reactivity:
- Mouse
- Mouse
- Host:
- Armenian hamster
- Armenian hamster
- Gene ID:
- 12501
- 12501
- Isotype:
- IgG
- IgG
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- +2 °C to +8 °C, protected from light
- +2 °C to +8 °C, protected from light