Order Entry
Czech Republic
ContactUsLinkComponent
 
Reversed phase HPLC columns, Synergi™ Polar-RP
Catalog # PHEN00D-4371-B0
Supplier:  PHENOMENEX
Reversed phase HPLC columns, Synergi™ Polar-RP
Catalog # PHEN00D-4371-B0
Supplier:  PHENOMENEX

Specifications

  • Column Format:
    Analytical
  • Ø int.:
    2,00 mm
  • Particle Size:
    2,5 µm
  • Pore size:
    100 Å
  • Length:
    100 mm
  • Seq:
    09
  • Pk:
    1

Specifications

About this item

Synergi™ Hydro-RP columns have polar endcapped, ether-linked phenyl phase providing polar and aromatic reversed phase selectivity.

  • Ideal for the separation of polar, aromatic compounds
  • Enhanced polar resolution in 100% buffer mobile phases
  • Highly reproducible and stable phenyl phase