Dissolution paddles, QLA
QLAAEPD016-02
- Description:Paddle, 419 mm, electropolished 316 stainless steel
- Seq:07
- Pk:1
QLA paddles are precision manufactured to limit rotational wobble to levels below USP (711) specifications.
- Made of high quality 316 electropolished stainless steel, PTFE coated over 316 stainless steel or solid fluoropolymer
- All paddles are laser marked with a unique serial number
The single piece molded Fluoropolymer paddles provide a seamless construction eliminating corrosion and contamination encountered with conventional three-piece assemblies.
: Other paddle shaft lengths are also available on request.
: All paddles are supplied with a Certificate of Conformance (COC) stating that they are USP compliant and that they meet machine manufacturer specifications. A Certificate of Analysis (COA) is available at an additional cost.