Test kits, DEHA, CHECKIT®
Catalog # LOVI147370
Supplier: Lovibond
CAS Number:
Specifications
- Pk:1 KIT
- Description:DEHA test kit
- Range:0 - 0,5 mg/l DEHA
Specifications
About this item
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
- Easy operation and exact reagent dosing
- Tablet reagents have minimum guaranteed shelf life of 5/10 years
The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.
Delivery: Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case.