Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tubing
Transfer Tubing
Tubing, PVC, with reinforcing
Tubing, PVC, with reinforcing
Tubing, PVC, with reinforcing
EDWAH02100017
:
EDWARDS HIGH VACUUM METTLER
Tubing, PVC, with reinforcing
EDWAH02100017
:
EDWARDS HIGH VACUUM METTLER
Tubing int.Ø:
25,4 mm
Description:
Reinforced PVC tube
Length:
1 m
Pk:
1