ContactUsLinkComponent
Weighing dishes, aluminium, WHEATON®
Weighing dishes, aluminium, WHEATON®
Catalog # WHEA370792
Supplier:  DWK Life Sciences
undefined
Weighing dishes, aluminium, WHEATON®
Catalog # WHEA370792
Supplier:  DWK Life Sciences
Specifications
  • Ext.Ø:
    20 mm
  • Height:
    8 mm
  • Pk:
    1.000
Specifications
About this item
Ideal for milligram weighing of liquids or solids.
  • Lightweight, easy to clean
  • Good chemical resistance
  • Good temperature resistance