ContactUsLinkComponent
Test kits, amine, Comparator 2000+
Test kits, amine, Comparator 2000+
Catalog # LOVI412740
Supplier:  Lovibond
CAS Number:  
undefined
Test kits, amine, Comparator 2000+
Catalog # LOVI412740
Supplier:  Lovibond
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Amines test kit
  • Seq:
    01
  • Type:
    AF 274
  • Range:
    1 - 10 mg/l
Specifications
About this item
Comparator 2000+ is a versatile modular system for testing water.
  • Stable and accurate
  • Robust, compact and portable
  • Prism enabling colour matching
  • Diffuser plate is used to measure the turbidity
  • Longer path length cells be used for lower concentration analysis
Delivery information: Test kit is supplied in a robust carrying case containing Comparator 2000+, amines comparator disc, extraction tube for result measurement, filter funnel and filter papers, 100 ml amber glass bottle, tablet reagents, crushing/stirring rod and instruction sheet.