

Calcium nitrate tetrahydrate 99-103% ACS
N261-1KG
:
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
- Barium:0.005%
- Chloride:= 0.005%
- Heavy Metals (as Pb):= 0.0005%
- Insolubles:= 0.005%
- Iron:= 0.0005%
- Magnesium:= 0.05%
- Nitrite:= 0.001%
- pH (5%, Water) @25°C:5 - 7
- Potassium:= 0.005%
- Purity:99% - 103%
- Sodium:= 0.01%
- Strontium:= 0.05%
- Sulphate:= 0.002%
:
:236.15 g/mol
:13,01687
:∼42 °C
:1.86 g/cm³ (20 °C)
:Ambient
:233-332-1
:1454
:5.1,III
:GHS07
:GHS05
:13477-34-4
:Ca(NO₃)₂·4H₂O
:MFCD00149604