Order Entry
Czech Republic
ContactUsLinkComponent
Test kits, photometric, phosphate by vanadate-molybdate method, LCK cuvette tests
Test kits, photometric, phosphate by vanadate-molybdate method, LCK cuvette tests
Catalog # 705-3062
Supplier:  Hach
CAS Number:  
undefined
Test kits, photometric, phosphate by vanadate-molybdate method, LCK cuvette tests
Catalog # 705-3062
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    Vanadato-molybdate
  • Pk:
    25 Tests
  • Description:
    Phosphate (ortho)
  • Type:
    LCK049
  • For unit:
    DR6000, DR3900, DR1900
  • Range:
    5 - 90 mg/l PO₄

Specifications

About this item

Cuvette tests for water testing applications.

  • Precise and reliable measurement values Maximum safety for users, thanks to the closed system and low amounts of reagents Convenient and error-free dosing of the reagents Barcode label for automatic recognition in the photometer Differentiation between tests and measuring ranges by means of colour-coding

Shelf life: 36 months from production date

Storage conditions: 15 to 25 °C

Accessories information: Cuvette tests are officially approved for many legally required consent limits, with the help of Addista standard solutions and round-robin test solutions, they provide the assurance you need.