In-Line filters, Polydisc™ HD, Whatman™
Polydisc HD (Heavy Duty) from Cytiva is a durable all-polypropylene in-line filter for large volumes of liquid samples containing even harsh chemical solvents.
- In-line, hose-connected filter with up to 1000 ml capacity
- High flow filter with durable all-polypropylene construction
- Broad aqueous and solvent compatibility
- 99% liquid retention rating
- Available in 5 and 10 µm pore size
- Recyclable after use
Labs that regularly process large sample volumes with different chemical compositions, pH values, and temperatures require a filter that is fast, durable, and broadly compatible.
Cytiva's Whatman brand heavy-duty Polydisc filter is a durable all-polypropylene filter. Its high strength, aqueous and solvent compatibility, and flow rate make it an excellent in-line filter for up to 1000 mL sample volumes
Frequently Bought Together