Order Entry
Czech Republic
ContactUsLinkComponent
Hydrazinium hydroxide ≥99% for synthesis
Toxic
Corrosive
Health
Environment
Hydrazinium hydroxide ≥99% for synthesis
Katalogové číslo MERC8.04608.0250
Supplier:  Merck
CAS Number:  
Toxic
Corrosive
Health
Environment
Hydrazinium hydroxide ≥99% for synthesis
Katalogové číslo MERC8.04608.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 ml
  • Seq:
    2590

Specifications

About this item

 :
 :50.06 g/mol
Boiling Pt:117.2 °C (1013 hPa)
Melting Pt:–51.7 °C
Density:1.03 g/cm³ (20 °C)
Flash Pt:75 °C
Storage Temperature:Ambient
EINECS:206-114-9
UN:2030
ADR:8 (6.1),II
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS05
 :GHS08
 :GHS09
CAS Number:7803-57-8
 :10217-52-4
Formula:NH₂NH₂·H₂O
MDL Number:MFCD00149931