Specifications
- Antibody type:Primary
- Antigen name:Vesicle-associated membrane protein 7
- Antigen symbol:VAMP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Gene ID:6845
- Antigen synonyms:SYBL1|TI-VAMP|TIVAMP|VAMP-7
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:E. coli-derived human SYBL1 recombinant protein (Position: N24-T190). Human SYBL1 shares 97.6% and 95.2% amino acid (aa) sequence identity with mouse and rat SYBL1, respectively.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Vesicle-associated membrane protein 7 (VAMP-7), also known as SYBL1, is a protein that in humans is encoded by the VAMP7 gene. This gene encodes a transmembrane protein that is a member of the soluble N-ethylmaleimide- sensitive factor attachment protein receptor (SNARE) family. The encoded protein localizes to late endosomes and lysosomes and is involved in the fusion of transport vesicles to their target membranes. Alternate splicing results in multiple transcript variants.
1. "Entrez Gene: SYBL1 synaptobrevin-like 1".
2. D'Esposito M, Ciccodicola A, Gianfrancesco F, Esposito T, Flagiello L, Mazzarella R, Schlessinger D, D'Urso M (Jul 1996). "A synaptobrevin-like gene in the Xq28 pseudoautosomal region undergoes X inactivation". Nat. Genet. 13 (2): 227–9.
3. Filippini F, Rossi V, Galli T, Budillon A, D'Urso M, D'Esposito M (Jul 2001). "Longins: a new evolutionary conserved VAMP family sharing a novel SNARE domain". Trends Biochem. Sci. 26 (7): 407–9.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.