Order Entry
Czech Republic
ContactUsLinkComponent
Anti-APOE Rabbit Polyclonal Antibody
Anti-APOE Rabbit Polyclonal Antibody
Catalog # BSBTPB9986
Supplier:  Boster Bio
undefined
Anti-APOE Rabbit Polyclonal Antibody
Catalog # BSBTPB9986
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    apolipoprotein E
  • Antigen symbol:
    APOE
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Gene ID:
    11816
  • Antigen synonyms:
    AI255918|Apo-E
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    E.coli-derived mouse Apolipoprotein E recombinant protein (Position: D55-Q294). Mouse Apolipoprotein E shares 75% and 94.6% amino acid (aa) sequence identity with human and rat Apolipoprotein E, respectively.
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

APOE is also known as AD2 or LPG. The protein encoded by this gene is a major apoprotein of the chylomicron. It binds to a specific liver and peripheral cell receptor, and is essential for the normal catabolism of triglyceride-rich lipoprotein constituents. This gene maps to chromosome 19 in a cluster with the related apolipoprotein C1 and C2 genes. Mutations in this gene result in familial dysbetalipoproteinemia, or type III hyperlipoproteinemia (HLP III), in which increased plasma cholesterol and triglycerides are the consequence of impaired clearance of chylomicron and VLDL remnants. Alternative splicing results in multiple transcript variants.

1. APOE ? variants increase risk of warfarin-related intracerebral hemorrhage.Falcone GJ, et al. Neurology, 2014 Sep 23.
2. Calabuig-Navarro MV, et al. Apolipoprotein E genotype has a modest impact on the postprandial plasma response to meals of varying fat composition in healthy men in a randomized controlled trial. J Nutr, 2014 Nov.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.