Beakers, glassy carbon
Katalogové číslo 39025.KT
Supplier: Thermo Fisher Scientific
Supplier #: 39025.KT
Log in for availability
About this item
Beakers, glassy carbon.
- Resistance to all wet decomposition agents
- No memory effects (uncontrolled adsorption and release of foreign elements)
- No contamination of analytical samples
- Stability to acid and alkaline melts
- No wetting effect of metal melts
- Good resistance to thermal shock allows rapid heating and cooling times
- Resistive to abrasive wear
- Good electrical conductivity
Vessels for ultra-high purity materials technology, e.g. semiconductor connections and crystalgrowing, e.g. doped halogenide crystals.
Frequently Bought Together