IPTG (isopropyl-β-D-thiogalactopyranoside), powder, Pierce™
Catalog # PIER34060
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:238.31 g/mol
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273