ContactUsLinkComponent
Oxidising additive, borer 44 PEROXYCLEAN
Oxidising additive, borer 44 PEROXYCLEAN
Catalog # DECO513705.00-K10M
Supplier:  Borer Chemie
undefined
Oxidising additive, borer 44 PEROXYCLEAN
Catalog # DECO513705.00-K10M
Supplier:  Borer Chemie
Specifications
  • Description:
    borer 44 PEROXYCLEAN
  • Pk:
    10 kg
Specifications
About this item
Cleaning booster with oxidising effect. For use in combination with alkaline or acidic deconex® cleaner.
  • In combination with deconex® cleaner, residue-free, rinsable and guarantees consistent and error-free results
  • Strongly oxidising additive for the automated cleaning of particularly stubborn residues
  • Perfect results even at low concentrations
  • Highly suitable for the effective cleaning of animal cages
  • Phosphate- and surfactant-free, no release of phosphates and surfactants into waste water
  • EDTA-, NTA- and chlorine-free, environmentally friendly