

2',3'-Didehydro-3'-deoxythymidine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3056-17-5
:
Formula:C₁₀H₁₂N₂O₄
MDL Number:MFCD00132921
:224.22 g/mol
Melting Pt:162…169 °C
:GHS07
:GHS08
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:160 to 169 °C
- HPLC:≥97.5 %
- Specific optical rotation:-44.0° to -48.0° (20 °C, 589 nm) (c=0.7, water)
Specification Test Results
Frequently Bought Together