

3-(Trifluoromethyl)piperidine 98%
Catalog # ACRO454820010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to light brown Liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
Specification Test Results
About this item
:
:153.15 g/mol
Boiling Pt:128…130 °C
Flash Pt:28 °C
Storage Temperature:Refrigerator
UN:1992
ADR:3,III
:GHS02
:GHS06
CAS Number:768-31-0
Formula:C₆H₁₀F₃N
MDL Number:MFCD02183561