Order Entry
Czech Republic
ContactUsLinkComponent
2-(Trifluoromethyl)pyrrolidine 97%
 
 
2-(Trifluoromethyl)pyrrolidine 97%
  ACRO454810050
 :  
undefined
 
 
2-(Trifluoromethyl)pyrrolidine 97%
  ACRO454810050
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Clear colorless to orange to light brown Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %
  • NMR (1H-NMR):
    Consistent with structure

 

 

 :
 :139.12 g/mol
 :110…112 °C
 :1.21 g/cm³
 :26 °C
 :1993
 :3,III
 :GHS02
 :GHS07
 :Refrigerator
 :109074-67-1
 :C₅H₈F₃N
 :MFCD02663405