
2,4-Dichloro-5-(trifluoromethyl)pyrimidine 97%
Catalog # ACRO454780010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to light brown Liquid or low melting solid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:216.98 g/mol
Boiling Pt:48…50 °C (32 mmHg)
Density:1.609 g/cm³
Flash Pt:93 °C
Storage Temperature:Freezer
UN:2810
ADR:6.1,III
:GHS06
CAS Number:3932-97-6
Formula:C₅HCl₂F₃N₂
MDL Number:MFCD03426408