ContactUsLinkComponent
2-Aminomethyl-1H-imidazole dihydrochloride 98%
Irritant
2-Aminomethyl-1H-imidazole dihydrochloride 98%
Certificates
undefined
Irritant
2-Aminomethyl-1H-imidazole dihydrochloride 98%
Specification Test Results
  • Appearance:
    White to light yellow to light brown Powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥97.5 to ≤102.5 %
  • HPLC:
    ≥97.5 %
  • NMR (1H-NMR):
    Consistent with structure