Order Entry
Czech Republic
ContactUsLinkComponent
Copper(II) bromide, anhydrous 99% for analysis
Irritant
Corrosive
Copper(II) bromide, anhydrous 99% for analysis
Certificates
undefined
Irritant
Corrosive
Copper(II) bromide, anhydrous 99% for analysis

Specification Test Results

  • Appearance:
    Silver grey to black Crystalline powder
  • Titration Iodometric:
    ≥98.5 %
  • Loss on drying:
    ≤1 %
  • Sulfate (SO4):
    ≤0.05 %
  • Iron (Fe):
    ≤0.01 %

Specification Test Results

Related Information

Frequently Bought Together