
1-Tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-amine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1190380-49-4
:
Formula:C₈H₁₃N₃O
MDL Number:MFCD11505007
:167.21 g/mol
Melting Pt:105…107 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow to pink Powder
- Infrared spectrum:Conforms
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together