
2-Chloro-4-methylpyrimidine 98%
Katalogové číslo ACRO453340100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:Colorless to white to light brown Low melting solid
- Infrared spectrum:Conforms
- GC:≥97.5 %
Specification Test Results
About this item
:
:128.56 g/mol
Boiling Pt:93…95 °C (17 mmHg)
Melting Pt:51…56 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:13036-57-2
Formula:C₅H₅ClN₂
MDL Number:MFCD00054434