Order Entry
Czech Republic
ContactUsLinkComponent
D-(-)-Fructose, meets analytical specification of USP
D-(-)-Fructose, meets analytical specification of USP
Certificates
undefined
D-(-)-Fructose, meets analytical specification of USP

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Assay:
    98.0 to 102.0 %
  • Loss on drying:
    ≤0.5 %
  • Residue after ignition:
    ≤0.5 %
  • Specific optical rotation:
    -91.0° to -93.5° (20°C, 589 nm)
  • Acidity:
    Passes test
  • Color of solution:
    Passes test
  • Chloride (Cl):
    ≤0.018 %
  • Sulfate (SO4):
    ≤0.025 %
  • Arsenic (As):
    ≤1 ppm
  • Ca and Mg:
    ≤0.005 % (as Ca)
  • Hydroxymethylfurfural:
    Passes test

Specification Test Results

Related Information

Frequently Bought Together