L-Aspartic acid monosodium salt monohydrate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:323194-76-9
:
Formula:C₄H₆NNaO₄·H₂O
MDL Number:MFCD00152960
:173.11 g/mol (155.10 anhy)
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Water:8 to 12 %
- Specific optical rotation:+19.0° to +21.0° (20°C, 589 nm) (c=10, 2.4M HCl)
- Titration with HClO4:≥96.0 %
Specification Test Results
Frequently Bought Together