

5-(Chloromethyl)-1,3-dimethyl-1H-pyrazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:852227-86-2
:
Formula:C₆H₉ClN₂
MDL Number:MFCD07368502
Melting Pt:65…66 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:144.61 g/mol
:GHS07
:GHS05
Specification Test Results
- Appearance:White to yellow Low melting solid
- Infrared spectrum:Conforms
- Melting point:63 to 69 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together