
3-Bromo-5-fluoropicolinonitrile 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:950670-18-5
:
Formula:C₆H₂BrFN₂
MDL Number:MFCD16988825
:201 g/mol
Melting Pt:88…91 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to light yellow Powder or solid
- Infrared spectrum:Conforms
- Melting point:88 to 92 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together