


Hydrogen chloride 4->6 N in 2-propanol, technical
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
:36.46 g/mol
Density:0.909 g/cm³
Flash Pt:11 °C
UN:2920
ADR:8,I
:GHS07
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Solution
- Water:≤5 %
- Strength:4 to 6 N
Specification Test Results
Frequently Bought Together